Learn More
MSC PRO-SAFE Hazardous Materials Spill Kit, 95 gal. Drum, 117 Piece, 1 ea.
Supplier: MSC 43827641
This hazardous materials spill kit is designed for the effective containment and cleanup of spill incidents. Housed in a durable 95 gal. polyethylene drum, this kit provides a comprehensive selection of sorbent materials to manage hazardous chemical spills efficiently.
- Includes 117 pieces for thorough spill response
- Features 100 green hazmat pads
- Contains 10 pillows and 4 booms
- Includes a pair of safety goggles and a pair of gloves
- Comes with a disposal bag for convenient cleanup
- Constructed with a polyethylene container