Learn More
Electron Microscopy Sciences ProScope 0-10x Lens
SDP
Supplier:  Electron Microscopy Sciences 6835210
Encompass
A variable focus lens that can be used as a webcam, still or video camera lens, or manually adjusted to 10x magnification. Has 12 white LEDs that can be use.
Catalog No. 50-336-55