Learn More
Electron Microscopy Sciences ProScope EDU w/ 5Mp imager
SDP
Supplier:  Electron Microscopy Sciences 68350EDU
Encompass
Designed for every level of science education from from Elementary to University, the ProScope-EDU does it all. It features a 5Mp imager for super sharp images, a 10x to 300x zoom lens, adjustable LEDs, a finite adjustable stand for hands-free use, software for Windows or Mac to capture stills, video and time-lapse, and connects via USB. The ProScope-EDU also works on Chromebooks and Android.
Catalog No. 50-192-7360