Learn More
Cayman Chemical Prostaglandin E2, 363-24-6, 1 mg
SDP
Supplier:  Cayman Chemical 140101
Encompass_Preferred
Molecular Formula C20H32O5, Purity ≥98%, Formula Weight 352.5, Synonyms: Dinoprostone, PGE2; PGE2 is one of the primary COX products of arachidonic acid and one of the most widely investigated PGs. Its activity influences inflammation, fertility and parturition, gastric mucosal integrity, and immune modulation.{1585,1558,437,1330} The effects of PGE2 are transduced by at least four distinct receptors designated EP1, EP2, EP3, and EP4.{1422} Affinity constants (Kd) of PGE2 for these receptors range from 1-10 nM depending on the receptor subtype and tissue.
Catalog No. NC9166732