Learn More
Medchemexpress LLC Prostatic acid phosp 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP7817A100UG
Encompass_Preferred
Acid prostatic phosphatase (ACPP) is an enzyme that catalyzes the conversion of orthophosphate monoesters into alcohol and orthophosphate ACPP is regulated by androgens and secreted by prostate epithelial cells Prostatic acid phosphatase/ACPP Protein Human (HEK293 His) is the recombinant human-derived Prostatic acid phosphatase/ACPP protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-93806