Learn More
Medchemexpress LLC HY-111758 10mg Medchemexpress, PROTAC B-Raf degrader 1 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11175810MG
Encompass_Preferred
Medchemexpress, HY-111758 10mg PROTAC B-Raf degrader 1 CAS: PROTAC B-Raf degrader 1 (compound 2) is a proteolysis targeting chimera (PROTAC) for the degradation of B-Raf. With anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8951