Learn More
Medchemexpress LLC Protac Bcl2 degrader-1 | 2378801-85-3 | 99.8% | 941.84 | C45H45BrN6O10S | 1MG

Supplier: Medchemexpress LLC HY1258761MG
PROTAC Bcl2 degrader-1 is a cereblon-based proteolysis targeting chimera (PROTAC) designed to induce ubiquitin-proteasome mediated degradation of antiapoptotic Bcl-2 family proteins. It is intended for cellular research into targeted protein degradation, apoptosis pathways, and mechanism-of-action studies; manufacturer data provide bioactivity, solubility, purity, and storage guidance to support experimental planning.
- Cereblon-based PROTAC that recruits CRBN E3 ligase.
- Induces selective degradation of Bcl-2 and Mcl-1 in cell assays.
- Reported bioactivity: Bcl-2 DC50 3 μM, Bcl-2 IC50 4.94 μM, Mcl-1 IC50 11.81 μM.
- Molecular formula C45H45BrN6O10S and molecular weight 941.84.
- High purity (99.8%) suitable for research use.
- Soluble in DMSO (≈25 mg/mL) and compatible with common in vivo formulations.
- Store at -20°C under inert atmosphere; in solvent store at -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.