Learn More
Medchemexpress LLC Protac Bcl2 degrader-1 | 2378801-85-3 | 99.8% | 941.84 g/mol | C45H45BrN6O10S | 10MG

Supplier: Medchemexpress LLC HY12587610MG
PROTAC Bcl2 degrader-1 is a cereblon-based proteolysis targeting chimera designed to selectively induce degradation of Bcl-2 and to show activity against Mcl-1. It is intended for biochemical and cellular research, supplied as a light yellow to yellow solid with high chemical purity and characterized potency in degradation assays.
- Induces degradation of Bcl-2 and shows activity against Mcl-1.
- Reported Bcl-2 IC50 4.94 μM and DC50 3.0 μM.
- High chemical purity of 99.8%.
- Molecular weight approximately 941.8 g/mol.
- Supplied as a light yellow to yellow solid for research use.
- Recommended storage at -20°C under inert gas; solvent stability and handling details available in technical documents.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.