Learn More
Medchemexpress LLC PROTAC Bcl2 degrader-1 | 2378801-85-3 | 99.8% | 941.84 g/mol | C45H45BrN6O10S | 5MG

Supplier: Medchemexpress LLC HY1258765MG
PROTAC Bcl2 degrader-1 is a cereblon-based PROTAC that selectively induces degradation of anti-apoptotic proteins Bcl-2 and Mcl-1 by recruiting the CRBN E3 ligase. Supplied for research use only as a high-purity solid, it is intended for biochemical and cellular studies of targeted protein degradation and apoptosis.
- Potent degrader of Bcl-2 and Mcl-1 (Bcl-2 DC50 3 μM; IC50 4.94 μM; Mcl-1 IC50 11.81 μM).
- High purity for research use (99.8%).
- Soluble in DMSO up to 25 mg/mL; in vivo vehicle formulations reported.
- Well characterized molecular properties (MW 941.84 g/mol; formula C45H45BrN6O10S).
- Storage conditions provided to preserve stability (-80°C in solvent for up to 6 months; -20°C for short term).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.