Learn More
Medchemexpress LLC PROTAC BCR-ABL Degrader-1 | C43H40N10O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15341550MG
Encompass_Preferred
PROTAC BCR-ABL Degrader-1 (compound PROTAC 1) is a PROTAC featuring a 2-oxoethyl linker. It functions by inducing Bcr-Abl degradation through a ubiquitinproteasom-dependent mechanism. This compound has demonstrated antiproliferative activity against K562 cells and shows promise for cancer research.
  • PROTAC with a 2-oxoethyl linker.
  • Induces Bcr-Abl degradation in a ubiquitinproteasom-dependent manner.
  • Exhibits antiproliferative activity against K562 cells.
  • Potential to study cancer.
Catalog No. 50-003-73706