Learn More
Medchemexpress LLC PROTAC BET Degrader-10 | 1957234-97-7 | 99.39% | 783.29 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112718
Encompass_Preferred
PROTAC BET Degrader-10 (dBET37) is a potent BET protein BRD4 degrader, connected by ligands for Cereblon and BRD4, with a DC50 of 49 nM. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Potent BET protein BRD4 degrader
  • Connects ligands for Cereblon and BRD4
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-39105