Learn More
Medchemexpress LLC PROTAC BET degrader- 10mg | 3031993-61-7 | 10 MG

Supplier: Medchemexpress LLC HY11422910MG
PROTAC BET degrader-3 is a bifunctional proteolysis targeting chimera that recruits the von Hippel-Lindau (VHL) E3 ligase to BET-family bromodomains to induce targeted degradation of BET proteins. It is supplied as a research-grade powder for preclinical studies.
- Targets BET proteins via VHL-mediated degradation.
- Molecular formula C53H64N12O9S.
- Molecular weight 1045.22 g·mol⁻¹.
- Purity 99.11% (HPLC).
- Powder form; store at -20°C, shelf life 3 years.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.