Learn More
Medchemexpress LLC Protac BRD4 degrader-5-co-peg3-n3 | 2704602-92-4 | 99.7% | 1231.87 | C58H75ClN12O12S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY13373610MG
Encompass_Preferred
PROTAC BRD4 Degrader-5-CO-PEG3-N3 is a PROTAC-linker conjugate that combines a BRD4 degrader with a PEG-based linker and an azide functional group for bioconjugation. It is provided as a high-purity research reagent for targeted protein degradation studies and click-chemistry applications.
  • azide functional group enables CuAAC and SPAAC conjugation
  • high purity suitable for research use
  • molecular weight 1231.87 and formula C58H75ClN12O12S2
  • available in small milligram quantities, including 10 mg
  • designed for use in antibody conjugation and linker chemistry workflows
Catalog No. 50-002-01955