Learn More
Medchemexpress LLC Protac BRD4 degrader-9 | 2417370-42-2 | 98.2% | 1312.50 g/mol | C59H71F2N9O15S4 | 1 MG

Supplier: Medchemexpress LLC HY1386321MG
PROTAC BRD4 Degrader-9 is a proteolysis-targeting chimera (PROTAC) that induces selective degradation of BRD4 in cellular assays. Supplied as a solid research compound for in vitro use only; not for human or clinical use.
- Targets BRD4 in cellular assays
- Cas number: 2417370-42-2
- Molecular formula: C59H71F2N9O15S4
- Molecular weight: 1312.50 g/mol
- Purity: 98.2%
- Storage: powder at -20°C; in solvent: -80°C for 6 months, -20°C for 1 month
- Available in mg-scale quantities suitable for research use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.