Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protac BRD4 degrader-9 | 2417370-42-2 | 98.2% | 1312.50 g/mol | C59H71F2N9O15S4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1386325MG

Encompass_Preferred

Protac BRD4 degrader-9 is a BRD4-targeting PROTAC small-molecule degrader provided for research use in targeted protein degradation and epigenetic studies. It is supplied as a powder with recommended frozen storage and is intended for biochemical and cellular assays.

  • Target: BRD4.
  • Molecular formula: C59H71F2N9O15S4.
  • Molecular weight: 1312.50 g/mol.
  • Purity: 98.2%.
  • Recommended storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-01982


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.