Learn More
Medchemexpress LLC HY-129939 5mg Medchemexpress, PROTAC BRD4 ligand-1 CAS:2313230-51-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1299395MG
Encompass_Preferred
Medchemexpress, HY-129939 5mg PROTAC BRD4 ligand-1 CAS:2313230-51-0 PROTAC BRD4 ligand-1 is a potent BET inhibitor and a ligand for target BRD4 protein for PROTACT. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9820