Learn More
Medchemexpress LLC Protac BTK Degrader-1 | 2801715-13-7 | 98.87% | 749.86 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1479435MG
Encompass_Preferred
PROTAC BTK Degrader-1 is a potent, selective, and orally active PROTAC BTK degrader with IC50 values of 34.51 nM for BTK WT and 64.56 nM for BTK-481S. It effectively reduces BTK protein levels and suppresses tumor growth. This degrader is also a click chemistry reagent, featuring an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Potent and selective degrader
  • Orally active
  • Reduces protein levels
  • Suppresses tumor growth
  • Click chemistry reagent
Catalog No. 50-003-62831