Learn More
Medchemexpress LLC PROTAC CBP/P300 Degrader-1 | 2484739-48-0 | 99.97% | 893.98 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138536100MG
Encompass_Preferred
PROTAC CBP/P300 Degrader-1 is a potent PROTAC CBP/P300 degrader. It potently inhibited cell viability of multiple cancer cell lines and is for research use only.
  • Shows LNCaP prostate cancer cell viability inhibition (IC50=0.4 nM).
  • Induces degradation of P300 (≥ 80%) at 10 nM.
  • Potently inhibited cell viability of multiple cancer cell lines (IC50s ranging from 0.1-141.4 nM).
Catalog No. 50-003-50040