Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protac CBP/P300 degrader-1 | 2484739-48-0 | Not specified | >95.0% | C46H53F2N11O6 | 10MG
SDP

Supplier:  Medchemexpress LLC HY13853610MG

Encompass_Preferred

PROTAC CBP/P300 Degrader-1 is a small-molecule proteolysis-targeting chimera that selectively induces degradation of the epigenetic reader proteins CBP and p300. It has been reported to potently inhibit cell viability in multiple cancer cell lines and is provided for research use only.

  • Targets CBP and p300 epigenetic reader domain.
  • Degrades target proteins via PROTAC mechanism.
  • Reported to inhibit viability of multiple cancer cell lines.
  • Molecular formula C46H53F2N11O6; molecular weight 893.98.
  • High solubility in DMSO (173.33 mg/mL); in vivo suspended solubility 6.5 mg/mL.
  • Storage: in solvent -80°C up to 6 months, -20°C up to 1 month; keep sealed, away from moisture and light.
  • Intended for laboratory research use only; not for human administration.

Catalog No. 50-002-61556


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.