Learn More
Medchemexpress LLC Protac cbp/p300 degrader-1 | 2484739-48-0 | 98.3% | 893.98 | 50 MG
Supplier: Medchemexpress LLC HY13853650MG
PROTAC CBP/P300 Degrader-1 is a potent PROTAC CBP/P300 degrader. It potently inhibits cell viability of multiple cancer cell lines.
- Potent PROTAC CBP/P300 degrader
- Inhibits cell viability of multiple cancer cell lines
- Shows LNCaP prostate cancer cell viability inhibition (IC50=0.4 nM)
- Induces degradation of P300 (≥ 80%) at 10 nM
- Targets Cereblon