Learn More
Medchemexpress LLC PROTAC CDK2/9 Degrader-1 | 2408641-24-5 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13070950MG
Encompass_Preferred
PROTAC CDK2/9 Degrader-1 (Compound F3) is a potent dual degrader for CDK2 (DC50=62 nM) and CDK9 (DC50=33 nM). It suppresses prostate cancer PC-3 cell proliferation (IC50=0.12 μM) by effectively blocking the cell cycle in S and G2/M phases. This compound is a PROTAC that functions by tethering a CDK inhibitor with a Cereblon ligand.
  • Potent dual degrader for CDK2 and CDK9
  • Suppresses prostate cancer PC-3 cell proliferation
  • Functions by tethering a CDK inhibitor with a Cereblon ligand
  • Induces cell cycle blockage at the G2/M phase
  • Down-regulates Mcl-1 protein level
  • Effectively degrades CDK2/9 in various cell lines
  • Inhibits both CDK2 and CDK9
Catalog No. 50-003-39945