Learn More
Medchemexpress LLC PROTAC CRBN Degrader-1 | 2358775-70-7 | 98.8% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12884510MG
Encompass_Preferred
PROTAC CRBN Degrader-1 (Compound 14a) is a VHL-CRBN heterodimerization PROTAC degrader that achieves a selective degradation of CRBN by binding two E3 ubiquitin ligases (VHL and CRBN) to each other.
  • VHL ligand (S,R,S)-AHPC
  • CRBN ligand Pomalidomide-C2-NH2
  • Linker
Catalog No. 50-003-40019