Learn More
Medchemexpress LLC Protac CRBN degrader-1 | 2358775-70-7 | MFCD32197292 | 98.8% | 1061.25 g/mol | C53H72N8O13S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1288455MG
Encompass_Preferred
PROTAC CRBN Degrader-1 is a research-grade proteolysis-targeting chimera that induces selective degradation of cereblon (CRBN) by promoting heterodimerization with VHL, enabling studies of targeted protein degradation and ubiquitin-proteasome-mediated pathways. Supplied as a purified small-molecule for in vitro and cellular research; not for human or diagnostic use.
  • Selective CRBN degradation via VHL-CRBN heterodimerization.
  • High purity suitable for biochemical and cellular assays.
  • Available in small-milligram research quantities.
  • Molecular formula C53H72N8O13S and molecular weight 1061.25 g/mol.
  • Accompanied by product documentation including SDS and COA.
Catalog No. 50-002-62321