Learn More
Medchemexpress LLC PROTAC EED degrader-2 | 2639882-69-0 | 99.9% | C50H58FN11O6S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13061510MG
Encompass_Preferred
PROTAC EED degrader-2 is a VHL-based proteolysis-targeting chimera that binds EED, a subunit of the polycomb repressive complex 2 (PRC2), promoting its ubiquitination and degradation. The compound demonstrates potent PRC2 inhibition in biochemical and cellular assays and is supplied as a high-purity solid for laboratory research use only.
  • induces targeted degradation of EED in cellular assays
  • demonstrates potent PRC2 inhibition (pIC50 ≈ 8.11)
  • high chemical purity suitable for biochemical studies
  • solid, white to off-white form with defined storage recommendations
  • detailed structural and analytical data available from supplier records
Catalog No. 50-002-60855