Learn More
Medchemexpress LLC PROTAC eEF2K degrader-1 | 2458170-54-0 | 98.6% | 707.65 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY146324
Encompass_Preferred
PROTAC eEF2K degrader-1 (Compound 11l) is an eEF2K-Targeting PROTAC small molecule that induces apoptosis in MDA-MB-231 cells and mediates eEF2K degradation. It is intended for research use only.
  • eEF2K-Targeting PROTAC small molecule
  • Induces apoptosis in MDA-MB-231 cells
  • Mediates eEF2K degradation
  • Solid, white to off-white in color
  • Soluble in DMSO at 25 mg/mL
  • Targets CaMK III/eEF-2K
Catalog No. 50-003-63353