Learn More
Medchemexpress LLC Protac EGFR degrader 4 | 2882845-50-1 | 99.7% | 995.29 | C55H70N12O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1463495MG
Encompass_Preferred
PROTAC designed to selectively target and induce degradation of mutant forms of the epidermal growth factor receptor (EGFR). Demonstrates potent degradation of EGFRdel19 and EGFRL858R/T790M in cellular assays and inhibits growth of EGFR-driven cancer cell lines. Supplied as a high-purity research reagent with recommended solvent and storage conditions.
  • Targets mutant EGFR, inducing degradation of activating and resistance variants.
  • Demonstrated cellular potency with low-nanomolar DC50 values.
  • High reported purity suitable for biochemical and cellular studies.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic treatment.
  • Store protected from light and under inert atmosphere; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-23117