Learn More
Medchemexpress LLC PROTAC EGFR degrader 8 | 2925923-46-0 | C40H46CIN11O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1539015MG
Encompass_Preferred
PROTAC EGFR degrader 8 (T-184) is a PROTAC EGFR degrader. It degrades EGFR in HCC827 cells with a DC50 of 15.56 nM and inhibits the growth of H1975, PC-9, and HCC827 cells with IC50s of 7.72 nM, 121.9 nM, and 14.21 nM, respectively. It can be utilized for cancer research, particularly NSCLC.
  • Degrades EGFR in HCC827 cell with a DC50 of 15.56 nM
  • Inhibits H1975, PC-9, HCC827 cell growth with IC50s of 7.72 nM, 121.9 nM, 14.21 nM
  • Used for research of cancer, especially NSCLC
Catalog No. 50-003-76496