Learn More
Medchemexpress LLC Protac ER degrader-4 | 2361114-15-8 | 98.8% | C53H67F3N6O8S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13530910MG
Encompass_Preferred
PROTAC ER Degrader-4 is a von Hippel-Lindau (VHL)-based proteolysis-targeting chimera that binds and induces degradation of the estrogen receptor (ER) in biochemical and cellular assays. It exhibits potent ER binding (IC50 0.8 nM) and induces ER degradation in MCF-7 cells (IC50 0.3 nM); supplied as a high-purity research compound for mechanistic studies of ER biology.
  • VHL-based PROTAC mechanism of action.
  • Potent ER binding (IC50 0.8 nM) and degradation in MCF-7 cells (IC50 0.3 nM).
  • High purity suitable for biochemical and cell-based studies.
  • Available in small research quantities.
  • Recommended storage at -20°C under inert gas; long-term stability in solvent at -80°C.
Catalog No. 50-002-59302