Learn More
Medchemexpress LLC PROTAC ERRα Degrader-3 | 2306388-65-6 | 98.8% | 956.99 | 50 MG

Supplier: Medchemexpress LLC HY13918550MG
PROTAC ERRα Degrader-3 is a potent and selective ERRα degrader based on a von Hippel-Lindau ligand. It can specifically degrade ERRα protein by more than 80% at a concentration of 30 nM and is inactive against ERRβ and ERRγ proteins. It induces ERRα degradation dose-dependently with an efficacious dose as low as 3.0 nM at 4.0 h, also decreasing levels of downstream target genes like ATP5B, MCAD, and PDK4.
- Potent and selective ERRα degrader
- Based on von Hippel-Lindau ligand
- Specifically degrades ERRα protein by >80% at 30 nM
- Inactive against ERRβ and ERRγ proteins
- Induces ERRα degradation dose-dependently from 0.3 nM to 10 μM within 4 hours
- Decreases protein levels of ERRα downstream target genes (ATP5B, MCAD, PDK4)
- Exhibits an IC50 value of 12.67 nM for blocking ERRα with PGC-1α peptide interaction
- Induces approximately 96% protein degradation at 100 nM after 4.0 h treatment
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.