Learn More
Medchemexpress LLC Protac ERRα degrader-3 | 2306388-65-6 | 98.8% | 956.99 g·mol⁻¹ | C47H50F6N6O7S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1391855MG
Encompass_Preferred
PROTAC ERRα Degrader-3 is a small-molecule proteolysis-targeting chimera (PROTAC) that selectively induces ubiquitin-dependent degradation of estrogen-related receptor alpha (ERRα) in biochemical and cell-based assays. It is intended for research use to probe ERRα function and to study PROTAC-mediated target degradation; users should consult the batch COA and SDS for analytical and safety data.
  • Selectively degrades ERRα in cellular assays.
  • Enables studies of targeted protein degradation mechanisms.
  • Suitable for biochemical and cell-based research applications.
  • Provided with batch-specific purity and analytical data.
  • Stable when stored at -20 °C according to supplier guidance.
Catalog No. 50-002-21342