Learn More
Medchemexpress LLC HY-119932 1mg Medchemexpress, PROTAC FAK degrader 1 CAS:2301916-69-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1199321MG
Encompass_Preferred
Medchemexpress, HY-119932 1mg PROTAC FAK degrader 1 CAS:2301916-69-6 PROTAC FAK degrader 1 is a selective and potent focal adhesion kinase (Fak) degrader with an IC50 of 6.5 nM, DC50 of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9552