Learn More
Medchemexpress LLC Protac FAK degrader 1 | 2301916-69-6 | 99.3% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11993210MG
Encompass_Preferred
PROTAC FAK degrader 1 is a proteolysis-targeting chimera designed to selectively induce degradation of focal adhesion kinase (FAK) in cellular models, enabling studies of FAK-dependent signaling, migration, and invasion.
  • Selective FAK degradation with DC50 of 3.0 nM in PC3 cells.
  • High purity (99.25%) suitable for cellular assays.
  • Available in small research sample sizes for in vitro use.
  • Useful for investigating cancer cell migration, invasion, and signaling pathways.
Catalog No. 50-002-32562