Learn More
Medchemexpress LLC Protac FKBP degrader-3 | 2079056-43-0 | 99.5% | 1295.54 g/mol | C68H90N6O17S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1353455MG
Encompass_Preferred
PROTAC FKBP Degrader-3 is a targeted protein degrader designed to recruit the VHL E3 ligase to FKBP proteins, promoting polyubiquitination and proteasomal degradation. It is supplied as a solid, off-white to light-yellow research reagent characterized for in vitro activity and formulated for biochemical and cell-based experiments. Typical handling includes storage at -20°C under nitrogen and dissolution in DMSO for assays.
Catalog No. 50-002-22668