Learn More
Medchemexpress LLC PROTAC(H-PGDS)-7 | 2761281-50-7 | 99.4% | 742.78 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13997250MG
Encompass_Preferred
PROTAC(H-PGDS)-7 is a Hematopoietic prostaglandin D synthase (H-PGDS) molecular glue degrader. It exhibits a DC50 of 17.3 pM and demonstrates potent suppression of prostaglandin D2 (PGD2) production in KU812 cells.
  • Functions as a hematopoietic prostaglandin D synthase (H-PGDS) molecular glue degrader
  • DC50 of 17.3 pM
  • Potently suppresses prostaglandin D2 (PGD2) production in KU812 cells
Catalog No. 50-003-51070