Learn More
Medchemexpress LLC Protac IDO1 degrader-1 | 2488851-89-2 | 99.0% | C40H53BrFN9O13 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13191110MG
Encompass_Preferred
PROTAC IDO1 Degrader-1 is a bifunctional small molecule designed to induce selective degradation of indoleamine 2,3-dioxygenase 1 (IDO1) by recruiting Cereblon E3 ligase, resulting in ubiquitination and proteasome-mediated clearance. It is intended as a research reagent for in vitro and biochemical studies probing IDO1 function and degradation mechanisms.
  • induces targeted degradation of IDO1 via the ubiquitin-proteasome system.
  • employs cereblon E3 ligase recruitment for selective target engagement.
  • enables functional studies of IDO1 biology and pathway modulation.
  • supplied as a characterized small-molecule research reagent for laboratory use.
  • provided with reported molecular formula and purity for assay planning.
Catalog No. 50-002-60948