Learn More
Medchemexpress LLC PROTAC IRAK4 degrader-1 | 2360533-90-8 | 99.8% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1299661MG
Encompass_Preferred
PROTAC IRAK4 degrader-1 (compound I-210) is a Cereblon-based IRAK4 PROTAC, composed of an IRAK4 ligand, an E3 ligase ligand, and a PROTAC linker. It is for research use only.
  • Cereblon-based IRAK4 PROTAC
  • Composed of three distinct parts: IRAK4 ligand, E3 ligase ligand, and PROTAC linker
  • Demonstrates IRAK4 protein degradation in OCI-LY-10 cells at concentrations of 0.01, 0.1, and 1 μM
Catalog No. 50-003-40076