Learn More
Medchemexpress LLC PROTAC IRAK4 degrader-1 | 2360533-90-8 | 99.8% | 100 MG

Supplier: Medchemexpress LLC HY129966100MG
PROTAC IRAK4 degrader-1 is a Cereblon-based IRAK4 PROTAC. It is composed of an IRAK4 ligand, an E3 ligase ligand (Pomalidomide), and a PROTAC linker. This compound is for research use only, not for sale to patients.
- Cereblon-based IRAK4 PROTAC
- Results in significant degradation of IRAK4 protein in OCI-LY-10 cells at varying concentrations
- Molecular weight: 904.85
- Formula: C44H39F3N12O7
- Appearance: White to yellow solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.