Learn More
Medchemexpress LLC PROTAC K-Ras Degrader-1 | 2378258-52-5 | 99.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12952310MG
Encompass_Preferred
PROTAC K-Ras Degrader-1 (Compound 518) is a PROTAC (Proteolysis Targeting Chimeras) that effectively degrades K-Ras. It is designed to work based on a Cereblon E3 ligand and exhibits a degradation efficacy of ≥70% in SW1573 cells. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Effectively degrades K-Ras
  • Functions based on a Cereblon E3 ligand
  • Exhibits ≥70% degradation efficacy in SW1573 cells
  • Utilizes the intracellular ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-39417