Learn More
Medchemexpress LLC PROTAC KRAS G12C degrader-2 | 2378257-72-6 | C51H53ClFN9O7 | 25 MG

Supplier: Medchemexpress LLC 25MGHY153821
PROTAC KRAS G12C degrader-2 is a modulator of K-Ras protein hydrolysis. It is a bifunctional compound, which contains on one end a cereblon inhibitor of apoptosis proteins (IAP) and on the other end a moiety which binds KRAS. This product is for research use only.
- Purity: 98.45%
- Molecular weight: 958.47
- Formula: C51H53ClFN9O7
- CAS number: 2378257-72-6
- Appearance: Solid
- Color: Light yellow to yellow
- Target: Ras, PROTACs
- Pathway: GPCR/G protein, MAPK/ERK pathway, PROTAC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.