Learn More
Medchemexpress LLC HY-W017772 100mg Medchemexpress, PROTAC Linker 10 CAS:139115-92-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW017772100MG
Encompass_Preferred
Medchemexpress, HY-W017772 100mg PROTAC Linker 10 CAS:139115-92-7 PROTAC Linker 10 is a PROTAC linker, which refers to the alkyl/ether composition. PROTAC Linker 10 can be used in the synthesis of a series of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4637