Learn More
Medchemexpress LLC Protac Mcl1 degrader-1 | 2163793-38-0 | 98.1% | 909.84 g·mol⁻¹ | C45H45BrN6O8S | 10MG
SDP
Supplier:  Medchemexpress LLC HY12587710MG
Encompass_Preferred
PROTAC Mcl1 degrader-1 is a cereblon-recruiting proteolysis-targeting chimera (PROTAC) designed to induce ubiquitination and proteasomal degradation of the anti-apoptotic protein Mcl-1. Supplied for research use, it is intended for biochemical and cellular studies of targeted protein degradation and Mcl-1 biology.
  • Induces ubiquitination and proteasomal degradation of Mcl-1.
  • Uses a cereblon E3 ligase ligand to recruit degradation machinery.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO for compound handling and dosing.
  • Available in multiple small-scale pack sizes for research use.
  • Not for human or clinical use.
Catalog No. 50-002-37971