Learn More
Medchemexpress LLC Protac Mcl1 degrader-1 | 2163793-38-0 | 98.1% | 909.84 g/mol | C45H45BrN6O8S | 5MG
SDP
Supplier:  Medchemexpress LLC HY1258775MG
Encompass_Preferred
PROTAC Mcl1 degrader-1 is a cereblon-based proteolysis targeting chimera (PROTAC) that induces ubiquitination and proteasomal degradation of the anti-apoptotic protein Mcl-1. It is provided as a small-molecule research compound for in vitro and in vivo studies; reported properties include molecular formula C45H45BrN6O8S, molecular weight 909.84 g/mol, CAS 2163793-38-0, and a reported IC50 of 0.78 μM for Mcl-1 activity. Recommended solvents and in vivo formulations are provided in the manufacturer datasheet.
  • Induces cereblon-mediated degradation of Mcl-1.
  • Potent Mcl-1 activity with reported IC50 of 0.78 μM.
  • Molecular weight 909.84 g/mol and formula C45H45BrN6O8S.
  • High solubility in DMSO; documented in vitro and in vivo formulations.
  • Intended for biochemical and cell-based research applications.
Catalog No. 50-002-35025