Learn More
Medchemexpress LLC Protac MDM2 degrader-2 | 2249944-99-6 | MFCD32197289 | 96.0% | 1391.22 g/mol | C70H76Cl4N10O12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1288415MG
Encompass_Preferred
PROTAC MDM2 Degrader-2 is a bifunctional small molecule degrader designed to recruit an E3 ligase to MDM2, promoting ubiquitination and proteasomal degradation of MDM2. The compound leads to p53 stabilization and shows antiproliferative effects in cellular assays. It is supplied for research use with characterized purity, formula, and storage recommendations.
Catalog No. 50-002-62183