Learn More
Medchemexpress LLC PROTAC MDM2 Degrader-3 | 2249750-23-8 | 95.01% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12884210MG
Encompass_Preferred
PROTAC MDM2 Degrader-3 is an MDM2 degrader based on PROTAC technology. It is composed of a potent MDM2 inhibitor, a linker, and an MDM2 ligand for E3 ubiquitin ligase. This product is intended for research use only and not for sale to patients.
  • Developed using PROTAC technology
  • Composed of a potent MDM2 inhibitor
  • Includes a linker
  • Contains an MDM2 ligand for E3 ubiquitin ligase
Catalog No. 50-003-40859