Learn More
Medchemexpress LLC Protac PD-1/PD-L1 degrader-1 | 2447066-37-5 | 99.5% | C59H58ClN7O11 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13118310MG
Encompass_Preferred
PROTAC PD-1/PD-L1 degrader-1 is a cereblon-based proteolysis targeting chimera designed to disrupt PD-1/PD-L1 interaction and induce degradation of PD-L1 in cells. It demonstrates potent cellular activity and has been applied in co-culture models to restore immune responses via a lysosome-dependent mechanism.
  • High purity suitable for cellular and biochemical assays.
  • Potent cellular activity (IC50 39.2 nM).
  • Cereblon E3 ligase-mediated degradation mechanism.
  • Reduces PD-L1 protein levels in a lysosome-dependent manner.
  • Suitable for studies of immune checkpoint modulation and PD-L1 biology.
Catalog No. 50-002-60455