Learn More
Medchemexpress LLC HY-111844 1mg Medchemexpress, PROTAC RAR Degrader-1 CAS:1351169-27-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1118441MG
Encompass_Preferred
Medchemexpress, HY-111844 1mg PROTAC RAR Degrader-1 CAS:1351169-27-1 PROTAC RAR Degrader-1 comprises a cIAP1 ligand binding group, a linker and a RAR ligand binding group. PROTAC RAR Degrader-1 is an RAR degrader. Maximal RAR degradation at 30 μM concentration in HT1080 cells. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8978