Learn More
Medchemexpress LLC Protac SGK3 degrader-1 | 2381320-35-8 | 99.9% | 1157.38 | C57H73FN10O11S2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY12587810MG
Encompass_Preferred
PROTAC SGK3 degrader-1 is a VHL-ligand-based proteolysis targeting chimera designed to induce degradation of Serum/Glucocorticoid Regulated Kinase 3 (SGK3) for biochemical and cell-based studies. The compound is supplied in small research pack sizes and is characterized by high analytical purity with supporting RP-HPLC documentation.
  • VHL-ligand-based PROTAC design for targeted protein degradation.
  • Efficient degradation of SGK3 in cell-based assays.
  • High purity suitable for research applications.
  • Available in small pack sizes for assay development.
  • Supported by RP-HPLC analytical documentation.
  • Validated for use in cell viability and western blot analyses.
Catalog No. 50-002-36221