Learn More
Medchemexpress LLC Protac SGK3 degrader-1 | 2381320-35-8 | 99.9% | 1157.38 g/mol | C57H73FN10O11S2 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1258781MG
Encompass_Preferred
PROTAC SGK3 degrader-1 is a VHL-ligand-based PROTAC chemical probe developed to induce proteasomal degradation of SGK3 in cellular systems. It exhibits rapid, concentration-dependent activity and is supplied as a high-purity research reagent.
Catalog No. 50-002-37784