Learn More
Medchemexpress LLC Protac SGK3 degrader-1 (synonyms: SGK3-PROTAC1) | 2381320-35-8 | 99.9% | 1157.38 g·mol-1 | C57H73FN10O11S2 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1258785MG
Encompass_Preferred
PROTAC SGK3 degrader-1 is a VHL-ligand based proteolysis targeting chimera designed to induce ubiquitin-proteasome-mediated degradation of SGK3. Supplied as a solid for research use, it is characterized by analytical methods for identity and purity and is intended for cellular degradation studies, chemical biology, and mechanistic investigation of SGK3 signaling.
  • Induces degradation of SGK3 in cell-based assays.
  • Characterized by molecular formula C57H73FN10O11S2 and molecular weight 1157.38 g·mol-1.
  • High purity suitable for biochemical and cellular studies.
  • Supplied in small research pack sizes for laboratory use.
  • Analytical data such as NMR and HPLC provided to support identity and purity.
Catalog No. 50-002-38342