Learn More
Medchemexpress LLC Protac SOS1 degrader-1 | 00-00-0 | >98.0% | 1051.8 g/mol | C57H76ClF7N10O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY145737A5MG
Encompass_Preferred
PROTAC SOS1 degrader-1 is a research-use small-molecule PROTAC that induces targeted degradation of the SOS1 protein and shows potent SOS1 engagement with antiproliferative activity in KRAS-mutant cancer cell lines. Supplied as a powder in small research pack sizes; follow the manufacturer's stability and storage recommendations.
  • Potent SOS1 degradation in cells (low-nanomolar DC50).
  • Strong antiproliferative activity in KRAS-mutant cancer cell lines.
  • High purity suitable for research applications (>98.0%).
  • Available in small-mass research quantities for assay development.
  • Stable when stored under recommended frozen and refrigerated conditions.
Catalog No. 50-002-21570